missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse ALDH1 / ALDH1A1 / ALDC Protein

Product Code. 16087321
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16087321

Brand: enQuireBio™ QP8967ye1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) ALDH1 / ALDH1A1 / ALDC, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_038495.2
Gene ID (Entrez) 11668
Name ALDH1 / ALDH1A1 / ALDC Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Aldh1a1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Yeast
Protein Tag His
Sequence SSPAQPAVPA PLADLKIQHT KIFINNEWHN SVSGKKFPVL NPATEEVICH VEEGDKADVD KAVKAARQAF QIGSPWRTMD ASERGRLLNK LADLMERDRL LLATMEALNG GKVFANAYLS DLGGCIKALK YCAGWADKIH GQTIPSDGDI FTYTRREPIG VCGQIIPWNF PMLMFIWKIG PALSCGNTVV VKPAEQTPLT ALHLASLIKE AGFPPGVVNI VPGYGPTAGA AISSHMDVDK VAFTGSTQVG KLIKEAAGKS NLKRVTLELG GKSPCIVFAD ADLDIAVEFA HHGVFYHQGQ CCVAASRIFV EESVYDEFVK RSVERAKKYV LGNPLTPGIN QGPQIDKEQH DKILDLIESG KKEGAKLECG GGRWGNKGFF VQPTVFSNVT DEMRIAKEEI FGPVQQIMKF KSVDDVIKRA NNTTYGLAAG LFTKDLDKAI TVSSALQAGV VWVNCYMMLS AQCPFGGFKM SGNGRELGEH GLYEYTELKT VAMKISQKNS
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.