missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse AHSG Protein
A cDNA sequence encoding the AHSG was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10984-1mg
Additional Details : Weight : 0.01000kg
Specifications
AHSG Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
APQGTGLGFR ELACDDPEAE QVALLAVDYL NNHLLQGFKQ VLNQIDKVKV WSRRPFGVVY EMEVDTLETT CHALDPTPLA NCSVRQLTEH AVEGDCDFHI LKQDGQFRVM HTQCHSTPDS AEDVRKLCPR CPLLTPFNDT NVVHTVNTAL AAFNTQNNGT YFKLVEISRA QNVPLPVSTL VEFVIAATDC TAKEVTDPAK CNLLAEKQHG FCKANLMHNL GGEEVSVACK LFQTQPQPAN ANAVGPVPTA NAALPADPPA SVVVGPVVVP RGLSDHRTYH DLRHAFSPVA SVESASGETL HSPKVGQPGA AGPVSPMCPG RIRHFKIHHH HHH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
AHSG protein solution (0.25 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |