missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse AHSA1 His Protein

Product Code. 18329875 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
50 μg
Unit Size:
100µg
200µg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329875 200 μg 200µg
18328822 50 μg 50µg
18372237 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18329875 Supplier Novus Biologicals™ Supplier No. NBP318324200UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

2uM generated a 9-fold ATPase activation of 2uM HSP90 (His-tagged HSP90 beta) in 33mM Hepes pH7.2, 30mM NaCl, 5mM MgCl2, 1mM DTT, 1.5mM ATP in a 100ul reaction at 37 degrees C. (This is an enzyme-linked ATP regeneration assay tracking loss of NADH absorbance at 340nm.)
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE, Western Blot
Formulation 25mM Hepes buffer pH7.2, 100mM KoAc, 50% glycerol
Gene ID (Entrez) 10598
Molecular Weight (g/mol) M.W. Theoretical: 38 kDa
Quantity 200 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1
Gene Symbol AHSA1
Research Category Signal Transduction
Purity or Quality Grade >85%
Protein AHSA1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.