missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse AFP / alpha-fetoProtein

Product Code. 15926489
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 15926489

Brand: enQuireBio™ QP5619ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) AFP / alpha-fetoprotein, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_031449.3
Gene ID (Entrez) 11576
Name AFP / alpha-fetoProtein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Afp
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His
Sequence KALHENEFGI ASTLDSSQCV TEKNVLSIAT ITFTQFVPEA TEEEVNKMTS DVLAAMKKNS GDGCLESQLS VFLDEICHET ELSNKYGLSG CCSQSGVERH QCLLARKKTA PASVPPFQFP EPAESCKAHE ENRAVFMNRF IYEVSRRNPF MYAPAILSLA AQYDKVVLAC CKADNKEECF QTKRASIAKE LREGSMLNEH VCSVIRKFGS RNLQATTIIK LSQKLTEANF TEIQKLALDV AHIHEECCQG NSLECLQDGE KVMTYICSQQ NILSSKIAEC CKLPMIQLGF CIIHAENGVK PEGLSLNPSQ FLGDRNFAQF SSEEKIMFMA SFLHEYSRTH PNLPVSVILR IAKTYQEILE KCSQSGNLPG CQDNLEEELQ KHIEESQALS KQSCALYQTL GDYKLQNLFL IGYTRKAPQL TSAELIDLTG KMVSIASTCC QLSEEKWSGC GEGMADIFIG HLCIRNEASP VNSGISHCCN SSYSNRRLCI TSFLRDETYA PPPFSEDKFI FHKDLCQAQG KALQTMKQEL LINLVKQKPE LTEEQLAAVT ADFSGLLEKC CKAQDQEVCF TEEGPKLISK TRDALGV
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.