missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ADK Protein
A cDNA sequence encoding the ADK was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10506-100ug
Additional Details : Weight : 0.01000kg
Specifications
11534 | |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 100 pmol/min/ and is defined as the amount of enzyme that convert 1.0 pmole of adenosine to AMP per minute at pH 7.5 at 37C in a couple system with PK and LDH. | |
Mouse | |
His | |
The ADK protein solution (1 mg/ml) contains 20% glycerol, 20mM Tris-HCl buffer (pH 8.0), 1mM EDTA and 50mM NaCl. |
ADK Protein | |
Research Use Only | |
ADK | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAADEP KPKKLKVEAP QALSENVLFG MGNPLLDISA VVDKDFLDKY SLKPNDQILA EDKHKELFDE LVKKFKVEYH AGGSTQNSMK VAQWLIQEPH KAATFFGCIG IDKFGEILKR KAADAHVDAH YYEQNEQPTG TCAACITGGN RSLVANLAAA NCYKKEKHLD LERNWVLVEK ARVYYIAGFF LTVSPESVLK VARYAAENNR VFTLNLSAPF ISQFFKEALM DVMPYVDILF GNETEAATFA REQGFETKDI KEIAKKAQAL PKVNSKRQRT VIFTQGRDDT IVAAENDVTA FPVLDQNQEE IIDTNGAGDA FVGGFLSQLV SDKPLTECIR AGHYAASVII RRTGCTFPEK PDFH | |
Greater than 95.0% as determined by SDS-PAGE. |