missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Adiponectin/Acrp30 Protein

Product Code. 18325018
Change view
Click to view available options
:
50ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18325018 50ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325018 Supplier R&D Systems™ Supplier No. 5095AC050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Adiponectin/Acrp30 Protein is derived from NS0. The Recombinant Mouse Adiponectin/Acrp30 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 11450
Molecular Weight (g/mol) 25.8 kDa
Name Adiponectin/Acrp30
Quantity 50μg
Source Mouse myeloma cell line,NS0-derived mouse Adiponectin/Acrp30 protein Glu18-Asn247,with a C-terminal 6-His tag
Storage Requirements Do not freeze. 6 months from date of receipt, 2 to 8°C as supplied.
Structural Form Oligomer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.