missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Adipocyte enhancer-binding protein 1

Product Code. 16080311
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16080311

Brand: enQuireBio™ QP7706ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Adipocyte enhancer-binding protein 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001278786.1
Gene ID (Entrez) 11568
Name Adipocyte enhancer-binding protein 1
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Aebp1
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His-SUMO
Sequence QTVLTDDEIE EFLEGFLSEL ETQSPPREDD VEVQPLPEPT QRPRKSKAGG KQRADVEVPP EKNKDKEKKG KKDKGPKATK PLEGSTRPTK KPKEKPPKAT KKPKEKPPKA TKKPKEKPPK ATKKPKEKPP KATKRPSAGK KFSTVAPLET LDRLLPSPSN PSAQELPQKR DTPFPNAWQG QGEETQVEAK QPRPEPEEET EMPTLDYNDQ IEKEDYEDFE YIRRQKQPRP TPSRRRLWPE RPEEKTEEPE ERKEVEPPLK PLLPPDYGDS YVIPNYDDLD YYFPHPPPQK PDVGQEVDEE KEEMKKPKKE GSSPKEDTED KWTVEKNKDH KGPRKGEELE EEWAP
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.