missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse ADAMTS1 Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
The Recombinant Mouse ADAMTS1 Protein is derived from CHO. The Recombinant Mouse ADAMTS1 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in Sodium Acetate, CaCl 2 and NaCl. |
| Molecular Weight (g/mol) | 53 kDa |
| Name | ADAMTS1 |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse ADAMTS1 protein Phe254-Cys725,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Monomer. Recombinant Mouse ADAMTS1 is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction