missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Active Heparanase/HPSE Protein

Product Code. 18369607
Change view
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369607 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369607 Supplier R&D Systems™ Supplier No. 9788GH005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to release biotinylated heparan sulfate from Recombinant Human Syndecan-4 (Catalog # 2918-SD ). 20 ng rmHPSE digestion will result in >50% of OD reduction compared with the Negative Control.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6YGZ1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and E64.
Gene ID (Entrez) 10855
Molecular Weight (g/mol) M.W. (Observed): 48-56 kDa (50 kDa subunit) and 8-12 kDa (8 kDa subunit), reducing conditions; M.W. (theoretical): 43 kDa (50 kDa subunit) and 9 kDa (8 kDa subunit)
Quantity 5 μg
Source Mouse myeloma cell line, NS0-derived mouse Heparanase/HPSE protein Asp28-Ile535, with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Cross Reactivity Heparanase/HPSE
Species Mouse
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Heparanase/HPSE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.