missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Active Cathepsin C/DPPI Protein

Product Code. 18365018
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365018 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18365018 Supplier R&D Systems™ Supplier No. 2336CY010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Mouse Active Cathepsin C/DPPI Protein is derived from NS0. The Recombinant Mouse Active Cathepsin C/DPPI Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2μm filtered solution in MES, NaCl and Glycerol.
Gene ID (Entrez) 13032
Molecular Weight (g/mol) 24 kDa (Pro domain), 18 kDa (Light/beta chain), and 9 kDa (Heavy/ alpha chain)
Name Active Cathepsin C/DPPI
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived mouse Cathepsin C/DPPI protein proform (Asp25-Leu462 with a C-terminal 10-His tag) The proform was activated by Recombinant Human Cathepsin L (Catalog &Num; 952-CY ) and further purified.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Structural Form Active form
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.