missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant MERS-CoV 3CL Protease

Product Code. 18756106
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18756106

Brand: R&D Systems™ E719050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carrier Free Recombinant Protein

Recombinant MERS-CoV virus 3CL Protease is a cysteine protease that cleaves proteins with sequences including a LQ[S/A/G]. The cleavage occurs c-terminal to the glutamine residue. We recommend an initial MERS-CoV 3CL Protease concentration of 50-100 nM when using substrate S-720.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number K9N638
For Use With (Application) Bioactivity
Formulation 1.7 mg/ml (50 μM) in 50 mM HEPES pH 7.5, 150 mM NaCl, 1 mM EDTA, 1 mM DTT
Gene ID (Entrez) 43740578
Molecular Weight (g/mol) M.W.-theoretical: 33 kDa
Quantity 50 μg
Source E. coli-derived mers-cov 3 CL Protease protein Ser3248 - Gln3553 Contains the following substitutions: Val 3551 to Leu; Met 3552 to Phe
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 48 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Alias 3 CL Proteinase, 3 C-like main protease, 3 CL-Mpro, COVID-19, M Proteinase, Main Protease, Mpro
Conjugate Unconjugated
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein SARS-CoV-2 3 CL Protease
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.