missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque CCL20 / MIP-3 alpha Protein

A DNA sequence encoding the Macaca mulatta (Rhesus macaque) CCL20 / MIP-3 alpha, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5772-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15937659

  • £2480.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001028026.1
CCL20 / MIP-3 alpha Protein
Research Use Only
CCL20
Recombinant Protein
E. coli
ASNFDCCLRY TDRILHPKFI VGFTQQLANE TCDINAVVFH TKKGLSVCAN PKQTWVKLIV RRLSKKINKM
Greater than 90% as determined by SDS-PAGE.
574182
1 mg
Not determined.
Full Length of Mature Protein
Macaque
His-GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers