missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant M. tuberculosis Peptidoglycan-binding protein ArfA Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Peptidoglycan-binding protein ArfA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9625-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
WP_003404684.1 | |
200 μg | |
Not determined. | |
Full Length | |
M. tuberculosis | |
His | |
Tris-based buffer, 50% glycerol |
M. tuberculosis Peptidoglycan-binding protein ArfA Protein | |
Research Use Only | |
arfA | |
Recombinant Protein | |
Yeast | |
MASKAGLGQT PATTDARRTQ KFYRGSPGRP WLIGAVVIPL LIAAIGYGAF ERPQSVTGPT GVLPTLTPTS TRGASALSLS LLSISRSGNT VTLIGDFPDE AAKAALMTAL NGLLAPGVNV IDQIHVDPVV RSLDFSSAEP VFTASVPIPD FGLKVERDTV TLTGTAPSSE HKDAVKRAAT STWPDMKIVN NIEVTGQAPP GPPASGPCAD LQSAINAVTG GPIAFGNDGA SLIPADYEIL NRVADKLKAC PDARVTINGY TDNTGSEGIN IPLSAQRAKI VADYLVARGV AGDHIATVGL GSVNPIASNA TPEGRAKNRR VEIVVN | |
Greater than 90% as determined by SDS-PAGE. |