missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant M. tuberculosis INHA Protein

A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) INHA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7385-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048590

  • £1930.00 / 500µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_003407553.1
500 μg
Not determined.
Full Length
M. tuberculosis
His
Tris-based buffer, 50% glycerol
M. tuberculosis INHA Protein
Research Use Only
INHA
Recombinant Protein
Yeast
MTGLLDGKRI LVSGIITDSS IAFHIARVAQ EQGAQLVLTG FDRLRLIQRI TDRLPAKAPL LELDVQNEEH LASLAGRVTE AIGAGNKLDG VVHSIGFMPQ TGMGINPFFD APYADVSKGI HISAYSYASM AKALLPIMNP GGSIVGMDFD PSRAMPAYNW MTVAKSALES VNRFVAREAG KYGVRSNLVA AGPIRTLAMS AIVGGALGEE AGAQIQLLEE GWDQRAPIGW NMKDATPVAK TVCALLSDWL PATTGDIIYA DGGAHTQLL
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers