missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant M. tuberculosis INHA Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) INHA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7385-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
WP_003407553.1 | |
10 μg | |
Not determined. | |
Full Length | |
M. tuberculosis | |
His | |
Tris-based buffer, 50% glycerol |
M. tuberculosis INHA Protein | |
Research Use Only | |
INHA | |
Recombinant Protein | |
Yeast | |
MTGLLDGKRI LVSGIITDSS IAFHIARVAQ EQGAQLVLTG FDRLRLIQRI TDRLPAKAPL LELDVQNEEH LASLAGRVTE AIGAGNKLDG VVHSIGFMPQ TGMGINPFFD APYADVSKGI HISAYSYASM AKALLPIMNP GGSIVGMDFD PSRAMPAYNW MTVAKSALES VNRFVAREAG KYGVRSNLVA AGPIRTLAMS AIVGGALGEE AGAQIQLLEE GWDQRAPIGW NMKDATPVAK TVCALLSDWL PATTGDIIYA DGGAHTQLL | |
Greater than 90% as determined by SDS-PAGE. |