missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Concentration | LYOPH |
| For Use With (Application) | SDS-PAGE |
| Formulation | Lyophilized powder containing HEPES, pH 7.5, NaCl, MgCl2, EDTA, DTT and a carbohydrate stabilizer. |
| Molecular Weight (g/mol) | M.W. Theoretical: 62 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20 degrees C. Avoid freeze/thaw cycles. |
| Gene Alias | firefly luciferase |
| Reconstitution | To obtain maximal solubility it is important to reconstitute the enzyme at a high salt concentration, such as 1 M Tris buffer with any counter ion at pH 7-8. Can be prepared at a concentration of up to 5 mg protein/ml. Do not vortex and avoid agitation. |
| Research Category | Cell Biology, Cellular Markers, Epitope Tags, Fusion Proteins |
| Purity or Quality Grade | >98%, by SDS-PAGE |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?