missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza Wisconsin 01/2010 Recombinant Protein
A cDNA sequence encoding the Wisconsin 01/2010 Recombinant was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13961-10ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza Wisconsin 01/2010 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant B/ Wisconsin 01/2010 solution contains 10mM Sodium phosphate, pH 7.1,150mM NaCl and 0.005% Tween-20. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |