missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza Matrix protein 1

Product Code. 16039101
Change view
Click to view available options
Quantity:
10 μg
100 μg
50 μg
Unit Size:
100µg
10µg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16039101 50 μg 50µg
16029101 10 μg 10µg
16019101 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16039101 Supplier enQuireBio™ Supplier No. QP9964iv50ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Influenza A virus (strain A/USA:Phila/1935 H1N1) Matrix protein 2, was expressed in the hosts and tags indicated.

Specifications

Name Influenza Matrix protein 1
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol M2
Protein Length Full Length Transmembrane Protein
Product Type Recombinant Protein
Cross Reactivity Influenza
Species E. coli
Protein Tag His
Sequence MSLLTEVETP IRNEWGCRCN GSSDPLVIAA SIIGILHLIL WILDRLLFKC IYRRFKYGLK RGPSTEGVPE SMREEYRKEQ QSAVDADDGH FVNIEPE
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.