missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza Massachusetts 02/2012 Recombinant Protein

Product Code. 15991099
Click to view available options
Quantity:
10 μg
100 μg
2 μg
Unit Size:
100µg
10µg
2µg
This item is not returnable. View return policy

Product Code. 15991099

Brand: enQuireBio™ QP12648100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Massachusetts 02/2012 Recombinant was constructed and used to recombinantly synthesize the protein.

Specifications

Name Influenza Massachusetts 02/2012 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Influenza
Species Baculovirus
Protein Tag Untagged
Buffer The Recombinant B/ Massachusetts 02/2012 solution contains 10mM Sodium phosphate, pH 7.1,150mM NaCl and 0.005% Tween-20.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.