missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H9N2 Hong-Kong Protein
A cDNA sequence encoding the H9N2 Hong-Kong was constructed and used to recombinantly synthesize the protein.
£99.15 - £1985.00
Specifications
Name | Influenza H9N2 Hong-Kong Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969358
|
enQuireBio™
QP12097-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959358
|
enQuireBio™
QP12097-10UG |
10 μg |
£219.00
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949358
|
enQuireBio™
QP12097-100UG |
100 μg |
£1985.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza H9N2 Hong-Kong Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant H9N2 A/Hong-Kong/1073/99 solution contains 10mM Sodium Phosphate, pH 7.0 and 150mM NaCl and 0.01% Tween-20. |