missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H3N2 Victoria Recombinant Protein
A cDNA sequence encoding the H3N2 Victoria Recombinant was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12090-2ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza H3N2 Victoria Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant H3N2 A/ Victoria 361/2011solution contains 10mM Sodium phosphate, pH 7.1,150mM NaCl and 0.005% Tween-20. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |