missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H3N2 Switzerland Recombinant Protein
A cDNA sequence encoding the H3N2 Switzerland Recombinant was constructed and used to recombinantly synthesize the protein.
£99.15 - £1985.00
Specifications
Name | Influenza H3N2 Switzerland Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15949328
|
enQuireBio™
QP12087-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939328
|
enQuireBio™
QP12087-10UG |
10 μg |
£219.00
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929328
|
enQuireBio™
QP12087-100UG |
100 μg |
£1985.00
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza H3N2 Switzerland Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant H3N2 A/ Switzerland 2013 solution contains 10mM Sodium phosphate, pH 7.1,150mM NaCl and 0.005% Tween-20. |