missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza H3N2 New-York Protein

A cDNA sequence encoding the H3N2 New-York was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12084-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15949318

  • £219.00 / 10µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Influenza H3N2 New-York Protein
Research Use Only
Recombinant Protein
Baculovirus
The Recombinant H3N2 A/New-York/55/04 solution contains 10mM Sodium phosphate, pH 7.2 and 150mM NaCl.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers