missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza-B Virus Florida 04/06 Protein
A cDNA sequence encoding the Influenza-B Virus Florida 04/06 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2205.00
Specifications
Name | Influenza-B Virus Florida 04/06 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15935459
|
enQuireBio™
QP14030-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955459
|
enQuireBio™
QP14030-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945459
|
enQuireBio™
QP14030-1MG |
1 mg |
£2205.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza-B Virus Florida 04/06 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined byAnalysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Influenza B virus | |
The B/Florida/04/06 solution contains STE, 0.1% sodium azide (NaN3) and 0.005% thimerosal. |