missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Influenza A Virus H3N2 Neuraminidase Protein
Description
Measured by its ability to cleave a fluorogenic substrate, 2’-(4-Methylumbelliferyl)-alpha-D-N-acetylneuraminic acid. The specific activity is >10,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | Q91MA2 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl2, DTT and Glycerol. |
| Molecular Weight (g/mol) | M.W. (Observed): 57-65 kDa, reducing conditions; M.W. (theoretical): 53 kDa |
| Name | Viral Neuraminidase |
| Quantity | 5 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived influenza a virus h3n2 Viral Neuraminidase protein Ala46-Ile469, vasodilator-stimulated phosphoprotein tetramerization domain and a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?