missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Influenza A Virus H1N1 Hemagglutinin Protein

Product Code. 18726436
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18726436

Brand: R&D Systems™ 11202HA100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

His-tag (Sf21-Expressed)

Measured by its binding ability in a functional ELISA. When Recombinant Human Galectin-1 (Catalog # 1152-GA ) immobilized at 10.00 μg/mL, 100 μL/well, the concentration of Recombinant Influenza A Virus H1N1 Hemagglutinin His-tag (Catalog # 11202-HA) that produces 50% of the optimal binding response is 0.500-5.00 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAD17229.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with Trehalose.
Molecular Weight (g/mol) M.W.-observed: 55-70 kDa, under reducing conditions., M.W.-theroretical: 58 kDa
Quantity 100 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived influenza a virus h1n1 Hemagglutinin protein Asp18-Gln529, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Hemagglutinin/HA Peptide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.