Learn More
Cytiva Recombinant Taq DNA Polymerase
Product Code. 11349951 Shop All Cytiva Products
Change view
Click to view available options
:
5000 units
Product Code.
11349951 5000 units
1 options
Product Code. 11349951 Supplier Cytiva Supplier No. 28937346
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For PCR and other procedures requiring DNA polymerase activity at elevated temperatures
Taq DNA Polymerase (cloned) is the recombinant form of the native enzyme from Thermus aquaticus expressed in E. coli.
  • Highly purified recombinant enzyme
  • Excellent batch-to-batch reproducibility
  • Each lot is tested to be free of nuclease contamination