missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zona pellucida sperm-binding protein 1

Product Code. 15982858
Click to view available options
Quantity:
100 μg
20 μg
50 μg
Unit Size:
100µg
20µg
50µg
This item is not returnable. View return policy

Product Code. 15982858

Brand: enQuireBio™ QP10107ma100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Zona pellucida sperm-binding protein 3, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001103824.1
Gene ID (Entrez) 7784
Name Zona pellucida sperm-binding protein 1
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ZP3
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species Mammal
Protein Tag His
Sequence QPLWLLQGGA SHPETSVQPV LVECQEATLM VMVSKDLFGT GKLIRAADLT LGPEACEPLV SMDTEDVVRF EVGLHECGNS MQVTDDALVY STFLLHDPRP VGNLSIVRTN RAEIPIECRY PRQGNVSSQA ILPTWLPFRT TVFSEEKLTF SLRLMEENWN AEKRSPTFHL GDAAHLQAEI HTGSHVPLRL FVDHCVATPT PDQNASPYHT IVDFHGCLVD GLTDASSAFK VPRPGPDTLQ FTVDVFHFAN DSRNMIYITC HLKVTLAEQD PDELNKACSF SKPSNSWFPV EGSADICQCC NKGDCGTPSH SRRQPHVMSQ WSRSASRNRR HVTEEADVTV GPLIFLDRRG DHEVEQWALP SDTSV
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.