missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZNF514 Protein
A cDNA sequence encoding the ZNF514 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP14004-1mg
Additional Details : Weight : 0.01000kg
Specifications
ZNF514 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTFEDVA VEFSQWEWGQ LNPAQKDLYR EVMLENFRNL AILGLLVSKP YVICQLEEGG EPFMVEREIS TGAHSDWKRR SKSKESMPSW GISKEELFQV VSVEKHIQDV LQFSKLKAAC GCDGQLEMQQ IKQERHLKQM STIHKSATTL SRDYKWNGFG RSLGLRSVLV NQHSILMGEG SYKCDTEFRQ TLGGNNSQRT HPEKKSCKCN ECGKSFHFQS ELRRHQRCHT GEKPYECSDC GRAFGHISSL IKHQRTHTGE KPYECSECGR AFSQSSSLVL HYRFHTGEKP YKCNECGRAF GHTSSLIKHQ RTHTGEKPYE CRECGRTFSQ SSSLIVHYRF HTGEKPYKCN KCGRAFSQSS SLTQHYRFHT GEKPYKCNEC GRAFAHTASL IKHQRSHAGK KTL | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ZNF514 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |