missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zinc finger matrin-type protein 1

A DNA sequence encoding the Homo sapiens (Human) Zinc finger matrin-type protein 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8082-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16082551

  • £384.00 / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001003692.1
Zinc finger matrin-type protein 1
Research Use Only
ZMAT5
Recombinant Protein
E. coli
MGKRYFCDYC DRSFQDNLHN RKKHLNGLQH LKAKKVWYDM FRDAAAILLD EQNKRPCRKF LLTGQCDFGS NCRFSHMSER DLQELSIQVE EERRAREWLL DAPELPEGHL EDWLEKRAKR LSSAPSSRAE PIRTTVFQYP VGWPPVQELP PSLRAPPPGG WPLQPRVQWG
Greater than 90% as determined by SDS-PAGE.
55954
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers