missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZC4H2 Protein
A cDNA sequence encoding the ZC4H2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13990-20ug
Additional Details : Weight : 0.01000kg
Specifications
ZC4H2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMADEQEI MCKLESIKEI RNKTLQMEKI KARLKAEFEA LESEERHLKE YKQEMDLLLQ EKMAHVEELR LIHADINVME NTIKQSENDL NKLLESTRRL HDEYKPLKEH VDALRMTLGL QRLPDLCEEE EKLSLDYFEK QKAEWQTEPQ EPPIPESLAA AAAAAQQLQV ARKQDTRQTA TFRQQPPPMK ACLSCHQQIH RNAPICPLCK AKSRSRNPKK PKRKQDE | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ZC4H2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl(pH 8.5), 50% glycerol, 0.2M NaCl and 2mM DTT. |