missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human YY1AP1 GST (N-Term) Protein

Código de producto. 18345829 Tienda Bio Techne Productos
Change view
Click to view available options
Quantity:
10 μg
25 μg
Tamaño de la unidad:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
18345829 10 μg 10µg
18379939 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18345829 Proveedor Novus Biologicals™ N.º de proveedor H00055249P0110ug

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Especificaciones

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 55249
Molecular Weight (g/mol) 107.03 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias FLJ10875, FLJ13914, HCCA1, HCCA2YY1APYAP, Hepatocellular carcinoma susceptibility protein, Hepatocellular carcinoma-associated protein 2, YY1 associated protein 1, YY1-associated protein 1
Gene Symbol YY1AP1
Research Category Cancer
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein YY1AP1
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.