missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human YEATS domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) YEATS domain-containing protein 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6895-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076610

  • £384.00 / 100µg
Estimated Shipment: 16-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001287879.1
YEATS domain-containing protein 1
Research Use Only
YEATS4
Recombinant Protein
E. coli
MFKRMAEFGP DSGGRVKGVT IVKPIVYGNV ARYFGKKREE DGHTHQWTVY VKPYRNEDMS AYVKKIQFKL HESYGNPLRV VTKPPYEITE TGWGEFEIII KIFFIDPNER PVTLYHLLKL FQSDTNAMLG KKTVVSEFYD EMIFQDPTAM MQQLLTTSRQ LTLGAYKHET EFAELEVKTR EKLEAAKKKT SFEIAELKER LKASRETINC LKNEIRKLEE DDQAKDI
Greater than 80% as determined by SDS-PAGE.
8089
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers