Learn More
R&D Systems™ Recombinant Human XIAP Western Blot Standard Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ WBC002
Additional Details : Weight : 0.00970kg
Product Code. 18307247
  • £131.75 / 100µL
Estimated Shipment: 03-06-2024
to see stock.
Beschreibung
Beschreibung
The Recombinant Human XIAP Western Blot Standard Protein has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
331
100μL
Unconjugated
XIAP Western Blot Standard
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Recombinant
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote