missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human XCL2/SCM-1 beta Protein

Product Code. 18726087
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18726087

Brand: R&D Systems™ 8418X2025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human XCL2/SCM-1 beta Protein is derived from HEK293. The Recombinant Human XCL2/SCM-1 beta Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UBD3
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
Gene ID (Entrez) 6846
Molecular Weight (g/mol) 10kDa
Name XCL2/SCM-1 beta
Preparation Method Reconstitute at 250μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Quality Control Testing SDS-PAGE: 19-22kDa, reducing conditions
Quantity 25 μg
Source Human embryonic kidney cell,HEK293-derived human XCL2/SCM-1 beta protein Val22-Gly114
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Val22
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 250 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.