missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Wnt-3a Protein
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and CHAPS.
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Unit Size:
10µg
500µg
Description
Measured by its ability to induce alkaline phosphatase production by MC3T3-E1 mouse preosteoblast cells. The ED50 for this effect is 5.00-25.0 ng/mL. Measured by its ability to induce Topflash reporter activity in HEK293T human embryonic kidney cells. The ED50 for this effect is <500 ng/mL. Protein concentrations should be titrated based on cell type and if appropriate, passage number of the cell line. Optimal concentrations should be determined by each laboratory for each application.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. |
| Gene ID (Entrez) | 89780 |
| Molecular Weight (g/mol) | 37.4 kDa |
| Name | Wnt-3a |
| Quantity | 500 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Wnt-3a protein Ser19-Lys352 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 75%, by SDS-PAGE with silver staining. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction