missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WIF1 Protein
Click to view available options
Quantity:
10 μg
2 μg
5 μg
Unit Size:
10µg
2µg
5µg
Specifications
Specifications
| Gene ID (Entrez) | 11197 |
| Name | WIF1 Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Gene Symbol | WIF1 |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | Baculovirus |
| Protein Tag | His |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction