missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VTA1 Protein

A cDNA sequence encoding the VTA1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13948-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955209

  • £155.00 / 20µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

VTA1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAALAP LPPLPAQFKS IQHHLRTAQE HDKRDPVVAY YCRLYAMQTG MKIDSKTPEC RKFLSKLMDQ LEALKKQLGD NEAITQEIVG CAHLENYALK MFLYADNEDR AGRFHKNMIK SFYTASLLID VITVFGELTD ENVKHRKYAR WKATYIHNCL KNGETPQAGP VGIEEDNDIE ENEDAGAASL PTQPTQPSSS STYDPSNMPS GNYTGIQIPP GAHAPANTPA EVPHSTGVAS NTIQPTPQTI PAIDPALFNT ISQGDVRLTP EDFARAQKYC KYAGSALQYE DVSTAVQNLQ KALKLLTTGR E
Greater than 95.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
VTA1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 10% glycerol and 200mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers