missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human VSTM2L His Protein
Shop All Bio Techne Products
Click to view available options
:
10μg; Unlabeled
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 25-204 of Human VSTM2L The Recombinant Human VSTM2L Protein is derived from E. coli. The Recombinant Human VSTM2L Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | Q96N03 |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. 20mM Tris-HCl buffer (pH8.0) containing 20% glycerol, 0.1M NaCl,1mM DTT, 0.1mM PMSF |
| Gene ID (Entrez) | 128434 |
| Molecular Weight (g/mol) | 22.6kDa |
| Name | VSTM2L Protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Purity or Quality Grade | >90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction