missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Vimentin Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Specifications
Specifications
| Accession Number | P08670, P08670, P08670 |
| For Use With (Application) | In vitro Assay, PAGE |
| Formulation | Lyophilized from a 0.2 um filtered solution in 30% Acetonitrile and 0.1 % TFA. |
| Gene ID (Entrez) | 7431 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 53.5 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of Vimentin as determined by LAL method. |
| Gene Alias | epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin |
| Gene Symbol | VIM |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction