missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Vesicle-associated membrane protein 1
A DNA sequence encoding the Homo sapiens (Human) Vesicle-associated membrane protein 7, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001138621.1 |
---|---|
Gene ID (Entrez) | 6845 |
Name | Vesicle-associated membrane protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036470
|
enQuireBio™
QP6876-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056470
|
enQuireBio™
QP6876-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046470
|
enQuireBio™
QP6876-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066470
|
enQuireBio™
QP6876-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076470
|
enQuireBio™
QP6876-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086470
|
enQuireBio™
QP6876-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001138621.1 | |
Vesicle-associated membrane protein 1 | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6845 | |
Research Use Only | |
VAMP7 | |
Recombinant Protein | |
E. coli | |
AILFAVVARG TTILAKHAWC GGNFLEVTEQ ILAKIPSENN KLTYSHGNYL FHYICQDRIV YLCITDDDFE RSRAFNFLNE IKKRFQTTYG SRAQTALPYA MNSEFSSVLA AQLKHHSENK GLDKVMETQA QVDELKGIMV RNIDLVAQRG ERLELLIDKT ENLVDSSVTF KTTSRNLARA MCMKN | |
Greater than 90% as determined by SDS-PAGE. |