missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Very long-chain acyl-CoA synthetase Protein

A DNA sequence encoding the Homo sapiens (Human) Very long-chain acyl-CoA synthetase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6694-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16025180

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001153101.1
Very long-chain acyl-CoA synthetase Protein
Research Use Only
SLC27A2
Recombinant Protein
Yeast
GATLALRTKF SASQFWDDCR KYNVTVIQYI GELLRYLCNS PQKPNDRDHK VRLALGNGLR GDVWRQFVKR FGDICIYEFY AATEGNIGFM NYARKVGAVG RVNYLQKKII TYDLIKYDVE KDEPVRDENG YCVRVPKGEV GLLVCKITQL TPFNGYAGAK AQTEKKKLRD VFKKGDLYFN SGDLLMVDHE NFIYFHDRVG DTFRWKGENV ATTEVADTVG LVDFVQEVNV YGVHVPDHEG RIGMASIKMK ENHEFDGKKL FQHIADYLPS YARPRFLRIQ DTIEITGTFK HRKMTLVEEG FNPAVIKDAL YFLDDTAKMY VPMTEDIYNA ISAKTLKL
Greater than 90% as determined by SDS-PAGE.
11001
50 μg
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers