missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human VEGF-B 167 Protein

Product Code. 18767336
Click to view available options
:
25 ug
This item is not returnable. View return policy

Product Code. 18767336

Brand: R&D Systems™ 751VEB025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Rat Neuropilin-1 Fc Chimera is immobilized at 1 μg/mL (100 μL/well), Recombinant Human VEGF-B 167 binds with an ED50 of 10 to 80 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAA91463
For Use With (Application) Bioactivity
Gene ID (Entrez) 7423
Molecular Weight (g/mol) 19 kDa (monomer)
Name vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL
Quantity 25 μg
Source E. coli-derived human VEGF-B protein Pro22-Arg188
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250 μg/mL in 4mM HCl.
Buffer Lyophilized from a 0.2 μm filtered solution in HCl with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.