missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human VEGF 165 Protein
Description
Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is 1.50-12.0 ng/mL.
Specifications
Specifications
| Accession Number | NP_001165097.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in Sodium Acetate. |
| Gene ID (Entrez) | 7422 |
| Molecular Weight (g/mol) | M.W. Observed: 19-21 kDa, under reducing conditions., M.W. Theoretical: 19.2 kDa (monomer) |
| Quantity | 50 μg |
| Source | E. coli-derived human VEGF protein Ala27-Arg191 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.01 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?