missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human VEGF 165 Protein

Product Code. 18117320
Click to view available options
Quantity:
1 mg
10 μg
50 μg
500 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
Unit Size:
10µg
1mg
500µg
50µg
This item is not returnable. View return policy

Product Code. 18117320

Brand: R&D Systems™ 293VE050/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is 1-6 ng/mL. The specific activity of recombinant human VEGF165 is approximately 1.7 x 103 U/μg, which is calibrated against recombinant human VEGF165 WHO Standard (NIBSC code: 02/286). Specific activity is for reference purposes only and is not routinely tested.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) observed_molecular_weight: 20-22 kDa, under reducing conditions. 39-42 kDa, under non-reducing conditions.
Name VEGF 165
Quantity 50 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human VEGF protein Ala27-Arg191
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS. Alternatively, reconstitute (at 100-500 μg/mL in sterile 4 mM HCl.)
Purity or Quality Grade 97%, by SDS-PAGE with silver staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.