missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human VEGF 145 (aa 27-171) Protein

Product Code. 18350108
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
Unit Size:
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18350108 Lyophilized from a 0.2μm filtered solution in HCl. 10µg
18310398 Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18350108 Supplier R&D Systems™ Supplier No. 7626VE010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human VEGF 145 (aa 27-171) Protein is derived from E. coli. The Recombinant Human VEGF 145 (aa 27-171) Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in HCl.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) 17 kDa (monomer)
Name VEGF 145 (aa 27-171)
Quantity 10 μg
Source E. coli-derived human VEGF protein Ala27-Arg171,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.