missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human VEGF 121 (aa 207-327) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 4644-VS-010
Additional Details : Weight : 0.00970kg
Description
Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is 1.00-8.00 ng/mL.Specifications
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. | |
14 kDa (monomer) | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
7422 | |
VEGF 121, aa 207-327 | |
E. coli-derived human VEGF protein Ala207-Arg327,with an N-terminal Met & Pro208-Arg327 | |
Disulfide-linked homodimer | |
Recombinant |