missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VAT1 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | VAT1 Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MSDEREVAEA ATGEDASSPP PKTEAASDPQ HPAASEGAAA AAASPPLLRC LVLTGFGGYD KVKLQSRPAA PPAPGPGQLT LRLRACGLNF ADLMARQGLY DRLPPLPVTP GMEGAGVVIA VGEGVSDRKA GDRVMVLNRS GMWQEEVTVP SVQTFLIPEA MTFEEAAALL VNYITAYMVL FDFGNLQPGH SVLVHMAAGG VGMAAVQLCR TVENVTVFGT ASASKHEALK ENGVTHPIDY HTTDYVDEIK KISPKGVDIV MDPLGGSDTA KGYNLLKPMG KVVTYGMANL LTGPKRNLMA LARTWWNQFS VTALQLLQAN RAVCGFHLGY LDGEVELVSG VVARLLALYN QGHIKPHIDS VWPFEKVADA MKQMQEKKNV GKVLLVPGPE KEN |
| Buffer | VAT1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 100mM NaCl. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction