missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Vasorin/SLIT-like 2 Fc Chimera Protein

Product Code. 18757006
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18757006

Brand: R&D Systems™ 10139VN050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vasorin, also known as SLIT-like 2, is an approximately 110-kDa transmembrane protein that is expressed on vascular smooth muscle cells and in the developing skeletal system

  • Recombinant Protein
  • Carrier Free (CF)
  • The carrier free version does not contain Bovine Serum Albumin (BSA)
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6EMK4
For Use With (Application) Bioactivity
Gene ID (Entrez) 114990
Molecular Weight (g/mol) 86 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human Vasorin/SLIT-like 2 protein

Human Vasorin/SLIT-like 2 (Cys24-Asn573) Accession # Q6EMK4IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.