missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Vascular endothelial growth factor D Protein

A DNA sequence encoding the Homo sapiens (Human) Vascular endothelial growth factor D, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8578-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16075381

  • £384.00 / 100µg
Estimated Shipment: 15-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004460.1
Vascular endothelial growth factor D Protein
Research Use Only
VEGFD
Recombinant Protein
E. coli
FAATFYDIET LKVIDEEWQR TQCSPRETCV EVASELGKST NTFFKPPCVN VFRCGGCCNE ESLICMNTST SYISKQLFEI SVPLTSVPEL VPVKVANHTG CKCLPTAPRH PYSIIRR
Greater than 90% as determined by SDS-PAGE.
2277
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers